Learn More
Medchemexpress LLC Glycolic acid oxidase inhibitor 1 | 77529-42-1 | 99.4% | 344.16 | 100 MG
SDP
Catalog No. 5000400712
Encompass_Preferred
Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1, in Table IV. It inhibits oxalate biosynthesis in isolated perfused rat liver.
  • It is a glycolate oxidase inhibitor.
  • It inhibits oxalate biosynthesis.
  • It provides a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.
Catalog No. 50-004-00712 Supplier Medchemexpress LLC Supplier No. HY22166100MG
Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1, in Table IV. It inhibits oxalate biosynthesis in isolated perfused rat liver.
  • It is a glycolate oxidase inhibitor.
  • It inhibits oxalate biosynthesis.
  • It provides a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.