Learn More
Medchemexpress LLC Glycolic acid oxidase inhibitor 1 | 77529-42-1 | 99.4% | 344.16 | 50 MG
SDP
Catalog No. 5000402617
Encompass_Preferred
Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1. It inhibits oxalate biosynthesis in isolated perfused rat liver and provides a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.
  • Inhibits oxalate biosynthesis in isolated perfused rat liver.
  • Provides a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.
Catalog No. 50-004-02617 Supplier Medchemexpress LLC Supplier No. HY2216650MG
Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1. It inhibits oxalate biosynthesis in isolated perfused rat liver and provides a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.
  • Inhibits oxalate biosynthesis in isolated perfused rat liver.
  • Provides a specific approach to the prevention and treatment of calcium oxalate renal lithiasis.