Learn More
Medchemexpress LLC Glycolic acid oxidase inhibitor 1 | 77529-42-1 | 99.4% | 344.16 | C16H10BrNO3 | 10 MG
SDP
Catalog No. 5000297588
Encompass_Preferred
Glycolic acid oxidase inhibitor 1 is a small-molecule glycolate oxidase inhibitor described in patent EP0021228A1 and supplied for laboratory research. It is reported to inhibit glycolate oxidase activity and reduce oxalate biosynthesis in perfused liver models; the compound is characterized by CAS 77529-42-1, molecular formula C16H10BrNO3, and molecular weight 344.16 g/mol.
  • Research use only.
  • Inhibits glycolate oxidase and reduces oxalate biosynthesis in biological models.
  • High reported purity (99.35%).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Characterized by CAS 77529-42-1, formula C16H10BrNO3, and molecular weight 344.16 g/mol.
Catalog No. 50-002-97588 Supplier Medchemexpress LLC Supplier No. HY2216610MG
Glycolic acid oxidase inhibitor 1 is a small-molecule glycolate oxidase inhibitor described in patent EP0021228A1 and supplied for laboratory research. It is reported to inhibit glycolate oxidase activity and reduce oxalate biosynthesis in perfused liver models; the compound is characterized by CAS 77529-42-1, molecular formula C16H10BrNO3, and molecular weight 344.16 g/mol.
  • Research use only.
  • Inhibits glycolate oxidase and reduces oxalate biosynthesis in biological models.
  • High reported purity (99.35%).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Characterized by CAS 77529-42-1, formula C16H10BrNO3, and molecular weight 344.16 g/mol.