Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycolic acid solution

Catalog No. 42060318L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
4 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-060-318L 18 L
42-060-31L 1 L
42-060-34L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 42-060-318L Supplier Sigma Aldrich Supplier No. 42060318L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycolic acid solution

Specifications

Melting Point 10°C
CAS 79-14-1
Boiling Point 112°C
Molecular Formula C2H4O3
Linear Formula HOCH2COOH
MDL Number MFCD00004312
Synonym Hydroxyacetic acid
Molecular Weight (g/mol) 76.05
Density 1.25 g/mL (at 25°C)
Quantity 18 L
Refractive Index n20/D 1.412
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.