Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycolic acid solution

Catalog No. 420581500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. 42-058-1500ML

Supplier: Sigma Aldrich 420581500ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycolic acid solution may be employed to evaluate the efficacy of glycolic peels treatment for all types of acne.DuPont product

Specifications

CAS 79-14-1
Boiling Point 112°C
Molecular Formula C2H4O3
Linear Formula HOCH2COOH
MDL Number MFCD00004312
Synonym Hydroxyacetic acid
Molecular Weight (g/mol) 76.05
Percent Purity 63-72% (by NaOH, titration)
Density 1.25 g/mL (at 25°C)
Quantity 500 mL
Refractive Index n20/D 1.408
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.