Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GlycoLink™ Coupling Catalyst

Catalog No. PI88944 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88944 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88944 Supplier Thermo Scientific™ Supplier No. 88944
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Enhance aldehyde-hydrazide crosslinking efficiency for glycoprotein immobilization and conjugation with analine (CAS 62-53-3) and an optimized buffer system.

The Thermo Scientific™ GlycoLink Coupling Catalyst decreases reaction times and increases aldehyde-hydrazide coupling efficiency, resulting in greater than 90% coupling of glycoproteins in 4 hours.

The GlycoLink Coupling Catalyst is a two-component kit composed of aniline and sodium acetate buffer. The catalyst accelerates the coupling rate of hydrazide and alkoxyamine moieties with reactive aldehydes that have been generated by periodate-oxidation of cis-diols in glycoproteins. The GlycoLink Coupling Catalyst increases the coupling efficiency of these reactions, decreasing the molar excess amounts of labeling reagents that are required for conjugation reactions. The catalyst is also useful for increasing the binding efficiency of oxidized glycoproteins or other carbohydrate-containing molecules to hydrazide resins.

Highlights:

  • Optimized – catalyzes aldehyde-hydrazide and aldehyde-alkoxyamine conjugation reaction

  • Efficient – maximizes coupling efficiency and decreases required coupling reaction incubation time

  • Gentle – does not interfere with protein function and is easily removed by desalting or dialysis

Recommended for:

Labeling aldehyde-containing molecules with alkoxyamine-biotins; Labeling aldehyde-containing molecules with hydrazide-biotins; Increase coupling efficiency of aldehyde-containing molecules to hydrazide resin

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Product Line GlycoLink
Form Liquid
Quantity 100 mL
Product Type Coupling Catalyst
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.