Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GlycoLink™ Immobilization Kit

Catalog No. PI88941 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 columns kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88941 10 columns kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88941 Supplier Thermo Scientific™ Supplier No. 88941
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Immobilize polyclonal antibodies or other glycoproteins, via periodate-oxidized sugars, onto hydrazide-activated polyacrylamide beads for AP, IP or pull-down assays.

Thermo Scientific™ GlycoLink Immobilization Kits provide affinity columns and reagents to immobilize glycoproteins through oxidized sugar groups for use in affinity-purification applications or studies of protein-protein interactions.

The GlycoLink Kits use hydrazide-activated resin and a special catalyst to prepare affinity columns with polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Any glycosylated proteins (even monoclonal antibodies) that contain periodate-oxidizable carbohydrate groups can be immobilized with these kits to make reusable affinity columns. Antibodies immobilized by this hydrazide immobilization chemistry have unobstructed antigen-binding sites and optimal purification capability. Prepared columns with stable glycoproteins or antibodies can be regenerated and reused at least five times for affinity-purification without significant loss in binding capacity.

Highlights:

  • High-capacity – immobilize 1 to 10mg of oxidized antibody or other glycoprotein per milliliter of resin

  • Fast and efficient – immobilize in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)

  • Specific immobilization – the hydrazide-activated UltraLink Resin conjugates only to purified glycoproteins whose sugar (e.g., sialic acid) groups have been gently oxidized with periodate

  • Stable attachment – resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilization step is required

  • Preserves antibody function – immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target

  • Convenient sizes – choose from two complete 10-column kits: one with 1mL columns and one with 0.1mL columns

UltraLink Hydrazide Resin:

  • Ligand: hydrazide
  • Support: Thermo Scientific UltraLink Biosupport
  • Spacer arm: 22 atoms
  • Loading: activated with at least 15μmol hydrazide per milliliter of settled resin
  • Format: 50% resin slurry in 0.02% sodium azide

Includes:

Kits contain GlycoLink Columns; coupling buffer, coupling catalyst and wash buffer; oxidizing reagent; desalting columns, PBS buffer pack; and column accessories

Recommended for:

Purify antigens using covalently coupled antibodies; Determine binding partners for a glycoprotein of interest; Perform large-scale affinity purifications

TRUSTED_SUSTAINABILITY

Specifications

Description GlycoLink Immobilization Kit
Content And Storage Sufficient For: Immobilizing 1 to 10 mg of glycoprotein per column
• GlycoLink Columns, 1 mL, 10 columns
• GlycoLink Coupling Buffer, 2 x 100 mL
• Aniline, 0.2 mL
• Wash Buffer, 120 mL
• Sodium meta-periodate, 0.5 g
• Zeba Spin Desalting Columns, 5 mL, 10 columns
• PBS Pack (makes 500mL), 1 pack
• Column Accessories, 10 ea

Store at 4°C.
Product Line GlycoLink
Quantity 10 columns kit
Type Immobilization Kit
Stationary Phase Hydrazide
Sufficient For 10 Columns
Format Spin Column
Column Type Affinity, UltraLink™ Resin
Binding Capacity 1 to 10 mg of GlycoProtein Immobilization/Column
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.