Learn More
Cayman Chemical GlycolIthocholIc ACD MaxSpec
SDP
Catalog No. 502988104
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
Catalog No. 50-298-8104 Supplier Cayman Chemical Supplier No. 31601100 UG
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications