Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolithocholic aci 100mg | 474-74-8 | 433.62 | C26H43NO4 | 100 MG
SDP

Catalog No. 5000228510
Encompass_Preferred

Glycolithocholic acid (lithocholylglycine) is an endogenous glycine-conjugated secondary bile acid supplied as an analytical standard for research and diagnostic assays, including studies of ulcerative colitis, non-alcoholic steatohepatitis, and primary sclerosing cholangitis.

  • High purity: 99.90%.
  • Molecular weight: 433.62 g/mol.
  • Chemical formula: C26H43NO4.
  • Available as solid packs including 100 mg, and smaller sizes.
  • Also available as a 10 mM solution in DMSO for ready use.
  • Includes supporting technical files: datasheet, COA, SDS, and spectral data.
  • Intended for analytical and research applications.

Catalog No. 50-002-28510 Supplier Medchemexpress LLC Supplier No. HY116374100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycolithocholic acid (lithocholylglycine) is an endogenous glycine-conjugated secondary bile acid supplied as an analytical standard for research and diagnostic assays, including studies of ulcerative colitis, non-alcoholic steatohepatitis, and primary sclerosing cholangitis.

  • High purity: 99.90%.
  • Molecular weight: 433.62 g/mol.
  • Chemical formula: C26H43NO4.
  • Available as solid packs including 100 mg, and smaller sizes.
  • Also available as a 10 mM solution in DMSO for ready use.
  • Includes supporting technical files: datasheet, COA, SDS, and spectral data.
  • Intended for analytical and research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.