Learn More
Medchemexpress LLC Glycolithocholic aci 10mg | 474-74-8 | 433.62 | C26H43NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11637410MG
Encompass_Preferred
Glycolithocholic acid is a glycine-conjugated secondary bile acid supplied as a high-purity analytical standard in solid form for research applications, including studies of bile acid metabolism and biomarker analysis in liver and gastrointestinal disease.
  • High purity: 99.90%.
  • Solid form, stable for storage and analytical use.
  • Molecular formula C26H43NO4 and molecular weight 433.62 g/mol.
  • Intended for use as an analytical standard in metabolomics and bile acid research.
  • Available in a 10 mg pack size for small-scale assays.
Catalog No. 50-003-08175