Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolithocholic aci 10mM 1mL | 24404-83-9 | 455.61 g/mol | C26H42NNaO4 | 1 ML
SDP

Catalog No. 5000227347
Encompass_Preferred

Glycolithocholic acid sodium is a glycine-conjugated secondary bile acid supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for use in biochemical and metabolic research focused on bile acid metabolism and signaling.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Purity 98.0%.
  • Molecular weight 455.61 g/mol.
  • Chemical formula C26H42NNaO4.
  • Suitable for studies of bile acid metabolism and signaling.
  • Also available as a solid in multiple sizes for custom preparation.

Catalog No. 50-002-27347 Supplier Medchemexpress LLC Supplier No. HY116374A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycolithocholic acid sodium is a glycine-conjugated secondary bile acid supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for use in biochemical and metabolic research focused on bile acid metabolism and signaling.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Purity 98.0%.
  • Molecular weight 455.61 g/mol.
  • Chemical formula C26H42NNaO4.
  • Suitable for studies of bile acid metabolism and signaling.
  • Also available as a solid in multiple sizes for custom preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.