Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolithocholic aci 25mg | 474-74-8 | 25 MG
SDP

Catalog No. 5000229877
Encompass_Preferred

Glycolithocholic acid is the glycine-conjugated form of the bile acid lithocholic acid, supplied as an analytical standard for studies of bile acid metabolism and endogenous metabolite analysis.

  • Glycine-conjugated bile acid used as an analytical standard and endogenous metabolite reference.
  • CAS 474-74-8.
  • Purity 99.90%.
  • Molecular formula C26H43NO4.
  • Molecular weight 433.62 g/mol.
  • Available in small sizes including 25 MG for analytical use.

Catalog No. 50-002-29877 Supplier Medchemexpress LLC Supplier No. HY116374R25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycolithocholic acid is the glycine-conjugated form of the bile acid lithocholic acid, supplied as an analytical standard for studies of bile acid metabolism and endogenous metabolite analysis.

  • Glycine-conjugated bile acid used as an analytical standard and endogenous metabolite reference.
  • CAS 474-74-8.
  • Purity 99.90%.
  • Molecular formula C26H43NO4.
  • Molecular weight 433.62 g/mol.
  • Available in small sizes including 25 MG for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.