Learn More
Medchemexpress LLC Glycolithocholic aci 25mg | 474-74-8 | 433.62 g/mol | C26H43NO4 | 25 MG
SDP
Catalog No. 5000228332
Encompass_Preferred
Glycolithocholic acid is a glycine-conjugated secondary bile acid and endogenous metabolite supplied as an analytical standard for research and diagnostic studies of liver and gastrointestinal disorders.
  • Used as an analytical standard for metabolite profiling and quantification.
  • Available as a solid and as 10 mM solution preparations for convenience.
  • High purity suitable for analytical and diagnostic applications.
  • Molecular formula C26H43NO4 and molecular weight 433.62 g/mol.
Catalog No. 50-002-28332 Supplier Medchemexpress LLC Supplier No. HY11637425MG
Glycolithocholic acid is a glycine-conjugated secondary bile acid and endogenous metabolite supplied as an analytical standard for research and diagnostic studies of liver and gastrointestinal disorders.
  • Used as an analytical standard for metabolite profiling and quantification.
  • Available as a solid and as 10 mM solution preparations for convenience.
  • High purity suitable for analytical and diagnostic applications.
  • Molecular formula C26H43NO4 and molecular weight 433.62 g/mol.