Learn More
Medchemexpress LLC Glycolithocholic aci 50mg | 474-74-8 | 433.62 | C26H43NO4 | 50 MG
SDP
Catalog No. 5000229928
Encompass_Preferred
Glycolithocholic acid is a glycine-conjugated secondary bile acid supplied as a solid analytical standard for research use. It is an endogenous metabolite used in studies and diagnostic research related to ulcerative colitis, non-alcoholic steatohepatitis, and primary sclerosing cholangitis.
  • Glycine-conjugated secondary bile acid.
  • Endogenous metabolite relevant to gastrointestinal and liver disease research.
  • Intended for research use only and analytical applications.
  • High purity: 99.90% as listed by manufacturer.
  • Supplied in milligram quantities for analytical workflows.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
Catalog No. 50-002-29928 Supplier Medchemexpress LLC Supplier No. HY11637450MG
Glycolithocholic acid is a glycine-conjugated secondary bile acid supplied as a solid analytical standard for research use. It is an endogenous metabolite used in studies and diagnostic research related to ulcerative colitis, non-alcoholic steatohepatitis, and primary sclerosing cholangitis.
  • Glycine-conjugated secondary bile acid.
  • Endogenous metabolite relevant to gastrointestinal and liver disease research.
  • Intended for research use only and analytical applications.
  • High purity: 99.90% as listed by manufacturer.
  • Supplied in milligram quantities for analytical workflows.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).