Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolithocholic aci 5mg | 24404-83-9 | 455.61 g/mol | C26H42NNaO4 | 5 MG
SDP

Catalog No. 5000308083
Encompass_Preferred

Glycolithocholic acid sodium (CAS 24404-83-9) is the glycine-conjugated sodium salt of lithocholic acid used as a research reagent and analytical standard in studies of bile acid metabolism and diagnostic biomarker research. It is supplied in small, assay-ready quantities and is available as both a solid and convenient DMSO solution for method development and quantitative assays.

  • Purity 98.0% suitable for analytical and research applications.
  • Available in small milligram pack sizes for standards and method validation.
  • Offered as solid and as 10 mM solution in DMSO for flexible use.
  • Molecular weight 455.61 g/mol and formula C26H42NNaO4 for accurate calculations.
  • White to off-white solid, convenient to handle and weigh for sample preparation.

Catalog No. 50-003-08083 Supplier Medchemexpress LLC Supplier No. HY116374A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycolithocholic acid sodium (CAS 24404-83-9) is the glycine-conjugated sodium salt of lithocholic acid used as a research reagent and analytical standard in studies of bile acid metabolism and diagnostic biomarker research. It is supplied in small, assay-ready quantities and is available as both a solid and convenient DMSO solution for method development and quantitative assays.

  • Purity 98.0% suitable for analytical and research applications.
  • Available in small milligram pack sizes for standards and method validation.
  • Offered as solid and as 10 mM solution in DMSO for flexible use.
  • Molecular weight 455.61 g/mol and formula C26H42NNaO4 for accurate calculations.
  • White to off-white solid, convenient to handle and weigh for sample preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.