Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolithocholic aci 5mg | 474-74-8 | 433.62 g·mol⁻¹ | C26H43NO4 | 5 MG
SDP

Catalog No. 5000228203
Encompass_Preferred

Glycolithocholic acid is a glycine-conjugated secondary bile acid and endogenous metabolite used as a research reagent and analytical standard for studies of bile acid metabolism and related diseases. It is applied in diagnostic and biochemical research, including investigations of ulcerative colitis, non-alcoholic steatohepatitis, and primary sclerosing cholangitis.

  • High purity analytical standard (99.90%).
  • Molecular formula C26H43NO4; molecular weight 433.62 g·mol⁻¹.
  • Available in small pack sizes suitable for analytical and research workflows.
  • Soluble in DMSO; recommended storage at -20 °C for long term.
  • Used in metabolic profiling, diagnostic assays, and bile acid research.

Catalog No. 50-002-28203 Supplier Medchemexpress LLC Supplier No. HY1163745MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycolithocholic acid is a glycine-conjugated secondary bile acid and endogenous metabolite used as a research reagent and analytical standard for studies of bile acid metabolism and related diseases. It is applied in diagnostic and biochemical research, including investigations of ulcerative colitis, non-alcoholic steatohepatitis, and primary sclerosing cholangitis.

  • High purity analytical standard (99.90%).
  • Molecular formula C26H43NO4; molecular weight 433.62 g·mol⁻¹.
  • Available in small pack sizes suitable for analytical and research workflows.
  • Soluble in DMSO; recommended storage at -20 °C for long term.
  • Used in metabolic profiling, diagnostic assays, and bile acid research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.