Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycolithocholic acid 3-sulfate disodium | 64936-82-9 | 99.7% | 557.65 | C26H41NNa2O7S | 10 MG
SDP

Catalog No. 5000284859
Encompass_Preferred

Glycolithocholic acid 3-sulfate disodium is a glycine-conjugated, sulfated secondary bile acid provided as a research reagent. It functions as a GPR39 agonist and has shown in vitro inhibition of HIV-1 replication, supporting its use in GPCR signaling, infection biology, and gallbladder disease studies.

  • GPR39 agonist activity observed in cell assays.
  • Inhibits HIV-1 replication in vitro.
  • Soluble in DMSO and formulatable for in vivo dosing.
  • Recommended sealed storage at -20°C; solvent stability guidelines available.
  • Molecular weight 557.65 and CAS 64936-82-9.

Catalog No. 50-002-84859 Supplier Medchemexpress LLC Supplier No. HYW58743010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycolithocholic acid 3-sulfate disodium is a glycine-conjugated, sulfated secondary bile acid provided as a research reagent. It functions as a GPR39 agonist and has shown in vitro inhibition of HIV-1 replication, supporting its use in GPCR signaling, infection biology, and gallbladder disease studies.

  • GPR39 agonist activity observed in cell assays.
  • Inhibits HIV-1 replication in vitro.
  • Soluble in DMSO and formulatable for in vivo dosing.
  • Recommended sealed storage at -20°C; solvent stability guidelines available.
  • Molecular weight 557.65 and CAS 64936-82-9.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.