Learn More
Cayman Chemical GlycolIthocholIc AcId-d4 500ug
SDP
Catalog No. 502991895
Encompass_Preferred
An internal standard for the quantification of glycolithocholic acid by GC- or LC-MS
Catalog No. 50-299-1895 Supplier Cayman Chemical Supplier No. 31308500 UG
An internal standard for the quantification of glycolithocholic acid by GC- or LC-MS