Learn More
TARGETMOL CHEMICALS INC Glycolithocholic acid sodium s
SDP
Catalog No. 502879066
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Glycolithocholic acid sodium salt, the sodium form of Glycolithocholic acid, is a glycine-conjugated secondary bile acid utilized in diagnosing ulcerative colitis (UC), non-alcoholic steatohepatitis (NASH), and primary sclerosing cholangitis (PSC). Purity 99.56%
Catalog No. 50-287-9066 Supplier TARGETMOL CHEMICALS INC Supplier No. T6433410MG
Add to Cart
Add to Cart
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Glycolithocholic acid sodium salt, the sodium form of Glycolithocholic acid, is a glycine-conjugated secondary bile acid utilized in diagnosing ulcerative colitis (UC), non-alcoholic steatohepatitis (NASH), and primary sclerosing cholangitis (PSC). Purity 99.56%