Learn More
Cayman Chemical GlycolIthocholIc AcIdodIM 10mg
SDP
Catalog No. 502984472
Encompass_Preferred
A glycine-conjugated form of lithocholic acid; increased in the liver of mice fed a diet supplemented with ursodeoxycholic acid; decreased in the plasma following subcutaneous administration of PEG-obestatin(Cys10, Cys13) in lean or diet-induced obese mice; serum levels increase with age in children
Catalog No. 50-298-4472 Supplier Cayman Chemical Supplier No. 2027310 MG
A glycine-conjugated form of lithocholic acid; increased in the liver of mice fed a diet supplemented with ursodeoxycholic acid; decreased in the plasma following subcutaneous administration of PEG-obestatin(Cys10, Cys13) in lean or diet-induced obese mice; serum levels increase with age in children