Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycoluril

Catalog No. G7305100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G7305100G 100 g
G73055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G7305100G Supplier Sigma Aldrich Supplier No. G7305100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycoluril

Specifications

CAS 496-46-8
Melting Point >300°C (lit.)
Molecular Formula C4H6N4O2
Linear Formula C4H6N4O2
MDL Number MFCD00005539
Quantity 100 g
Synonym Acetylene carbamide; Acetylenediurea; NSC 2765; Tetrahydroimidazo[4,5-d]imidazole-2,5-dione
Molecular Weight (g/mol) 142.12
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.