Learn More
Cell Signaling Technology Glycolysis Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502051235
Encompass
The Glycolysis Antibody Sampler Kit provides an economical solution for investigating select enzymes involved in glycolysis. This kit contains primary antibodies to perform western blot experiments to detect endogenous levels of total target proteins, including key glycolytic enzymes like hexokinase, phosphofructokinase, and pyruvate kinase, as well as the pyruvate dehydrogenase complex. The kit includes an HRP-linked anti-rabbit IgG antibody for detection.
  • Economical selection of antibodies for studying glycolytic pathways
  • Includes primary antibodies for GAPDH, LDHA, Hexokinase 1 and 2, PKM1/2, PKM2, PDHA1, and PFKP
  • Supplied with HRP-linked secondary antibody
  • Validated for western blot analysis
  • Contains 20 μL of each primary antibody
Catalog No. 50-205-1235 Supplier Cell Signaling Technology Supplier No. 8337T
The Glycolysis Antibody Sampler Kit provides an economical solution for investigating select enzymes involved in glycolysis. This kit contains primary antibodies to perform western blot experiments to detect endogenous levels of total target proteins, including key glycolytic enzymes like hexokinase, phosphofructokinase, and pyruvate kinase, as well as the pyruvate dehydrogenase complex. The kit includes an HRP-linked anti-rabbit IgG antibody for detection.
  • Economical selection of antibodies for studying glycolytic pathways
  • Includes primary antibodies for GAPDH, LDHA, Hexokinase 1 and 2, PKM1/2, PKM2, PDHA1, and PFKP
  • Supplied with HRP-linked secondary antibody
  • Validated for western blot analysis
  • Contains 20 μL of each primary antibody