Learn More
Dojindo Molecular Technologies Inc GLYCOLYSIS/OXPHOS ASSAY KT,50T
SDP
Catalog No. NC2261647
Encompass
Glycolysis/Oxidative phosphorylation AssayEasy test via plate reader, no need for expensive equipmentAll reagent acquired is included, ready to use kitEasy-to-understand detailed protocol
This item is not returnable. View return policy
Catalog No. NC2261647 Supplier Dojindo Molecular Technologies Inc Supplier No. G27010
Glycolysis/Oxidative phosphorylation AssayEasy test via plate reader, no need for expensive equipmentAll reagent acquired is included, ready to use kitEasy-to-understand detailed protocol