Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP7413EC10UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His-SUMO. Host: E. coli. Species Reactivity: RABV. Protein Expression: A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated. AA Sequence: See company website. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: G. Uniprot: P03524.
Catalog No. 50-188-7061