Learn More
Biotium Glycophorin A / CD235a(GPHN02), 0.2mg/mL
Supplier:  Biotium BNUB0865100
Encompass_Preferred
Recognizes a sialoglycoprotein of 39 kDa, identified as glycophorin A (GPA). It is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorin acts as the receptor for Sandei virus and parvovirus. Glycophorins A (GPA) and B (GPB), which are single, trans-membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation.This antibody is available purified with BSA/azide at 200 ug/mL, or BSA/azide-free at 1 mg/mL.
Catalog No. 50-252-2884