Learn More
Diagnostic Biosystems GLYCOPHORIN C Glycophorin C
Catalog No. 502407493
Encompass_Preferred
This antibody is specific to β sialoglycophorin also known as glycophorin C. Glycophorin C is found on the human erythrocyte membrane and is one of four sialic acid-rich polypeptides.
Catalog No. 50-240-7493 Supplier Diagnostic Biosystems Supplier No. PDM578
This antibody is specific to β sialoglycophorin also known as glycophorin C. Glycophorin C is found on the human erythrocyte membrane and is one of four sialic acid-rich polypeptides.