Learn More
Bioss Glycophorin C/CD236 Recombinan
SDP
Catalog No. 5000065395
Encompass_Preferred
Glycophorin C/CD236 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65395 Supplier Bioss Supplier No. BSM61751R
Glycophorin C/CD236 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol