Learn More
Sigma Aldrich Fine Chemicals Biosciences GlycoProfile™ 2-AB Labeling Kit
Catalog No. NC1810037
Encompass
The GlycoProfile™ labeling kits are designed for efficient labeling of N-linked, O-linked, and glycosyl­phosphatidyl­inositol (GPI) anchored glycans with either 2-aminobenzamide (2-AB) or 2-aminobenzoic acid (anthranilic acid; 2-AA). These small fluorophores increase the spectral absorption of glycans, improving the detection of labeled glycans by HPAE (high-performance anion exchange chromatography), HPLC, and ESI-MS (electrospray mass spectroscopy) methods. Binding is robust and the resulting dye-glycan conjugates are stable, with no degradation during post-labeling analysis.
Catalog No. NC1810037 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PP05201KT
The GlycoProfile™ labeling kits are designed for efficient labeling of N-linked, O-linked, and glycosyl­phosphatidyl­inositol (GPI) anchored glycans with either 2-aminobenzamide (2-AB) or 2-aminobenzoic acid (anthranilic acid; 2-AA). These small fluorophores increase the spectral absorption of glycans, improving the detection of labeled glycans by HPAE (high-performance anion exchange chromatography), HPLC, and ESI-MS (electrospray mass spectroscopy) methods. Binding is robust and the resulting dye-glycan conjugates are stable, with no degradation during post-labeling analysis.