Learn More
ACROBiosystems Rabies virus Glycoprotein (strain CVS-11), His Tag (RAG-V55H5) is expressed from InsectCells-SF9.
Catalog No. 502231066
Encompass_Preferred
Rabies virus (RABV), scientific name Rabies lyssavirus, is a deadly neurotropic virus that causes rabies in humans and animals. Rabies virus has an extremely wide host range and its transmission most often occur through the saliva of animals. Without intervention prior to disease progression, rabies has the highest case fatality of any infectious disease. RABV contains a single-stranded negative-sense RNA genome that encodes five structural proteins: nucleoprotein (N), phosphoprotein (P), matrix protein (M), glycoprotein (G), and RNA-dependent RNA polymerase (L). Among these viral proteins, the RABV glycoprotein (RABV-G) is a pivotal player mediating virus entry and the major target of neutralizing antibodies, thus a key factor for vaccine and drug design.
Catalog No. 50-223-1066 Supplier ACROBiosystems Supplier No. RAGV55H550UG
Rabies virus (RABV), scientific name Rabies lyssavirus, is a deadly neurotropic virus that causes rabies in humans and animals. Rabies virus has an extremely wide host range and its transmission most often occur through the saliva of animals. Without intervention prior to disease progression, rabies has the highest case fatality of any infectious disease. RABV contains a single-stranded negative-sense RNA genome that encodes five structural proteins: nucleoprotein (N), phosphoprotein (P), matrix protein (M), glycoprotein (G), and RNA-dependent RNA polymerase (L). Among these viral proteins, the RABV glycoprotein (RABV-G) is a pivotal player mediating virus entry and the major target of neutralizing antibodies, thus a key factor for vaccine and drug design.