Learn More
QA Bio Inc QA-Bio, Inc. • LudgerTag DMB Sialic Acid Labeling Kit
Catalog No. 50355849
Encompass
DMB Sialic Acid Labeling Kit • The LudgerTag DMB Sialic Acid Labeling Kit has been developed for the analysis of sialic acids. Includes reagents for the release of sialic acids, DMB labeling reagents, a sialic acid reference panel, and the NANA and NGNA standards.
Catalog No. 50-355-849 Supplier QA Bio Inc Supplier No. LTKDMBA1
DMB Sialic Acid Labeling Kit • The LudgerTag DMB Sialic Acid Labeling Kit has been developed for the analysis of sialic acids. Includes reagents for the release of sialic acids, DMB labeling reagents, a sialic acid reference panel, and the NANA and NGNA standards.