Learn More
QA Bio Inc QA-Bio, Inc. • LudgerClean EB10 Glycan Cleanup Cartridges • 6 cartridges
Catalog No. 50355831
Encompass
EB10 Glycan Cleanup Cartridges • Ludger EB10 cartidges have been designed for purification of glycans from proteins, salts, and detergents.Applications include cleanup of glycans following hydrazinolysis, endoglycosidase digests, and enzyme treatment.
Catalog No. 50-355-831 Supplier QA Bio Inc Supplier No. LCEB10A6
EB10 Glycan Cleanup Cartridges • Ludger EB10 cartidges have been designed for purification of glycans from proteins, salts, and detergents.Applications include cleanup of glycans following hydrazinolysis, endoglycosidase digests, and enzyme treatment.