Learn More
QA Bio Inc QA-Bio, Inc. • LudgerSep N Buffer for Glycan Analysis • Normal Phase Buffer
Catalog No. 50355838
Encompass
LudgerSep N Buffer for Glycan Analysis • For preparation of LS-N Buffer (50 mM ammonium formate buffer, pH 4.4) used in amide or HILIC HPLC analysis of LudgerTag fluorophore and UV-chromophore labeled glycans.
Catalog No. 50-355-838 Supplier QA Bio Inc Supplier No. LSNBUFFX40
LudgerSep N Buffer for Glycan Analysis • For preparation of LS-N Buffer (50 mM ammonium formate buffer, pH 4.4) used in amide or HILIC HPLC analysis of LudgerTag fluorophore and UV-chromophore labeled glycans.