Learn More
QA Bio Inc QA-Bio, Inc. • LudgerClean S Cartridges • Glycan Cleanup after Fluorescent Labeling • 6 cartridges
Catalog No. 50355833
Encompass
S Cartridges -Fluorescent Labeling Cleanup for Glycans• One of the most reliable methods for achieving post-labeling cleanup is use of S-cartridges. Ludger S Cartridges are routinely used in the biopharma industry during glycoprofiling QC for lot release of FDA and EMA approved biopharmaceuticals.
Catalog No. 50-355-833 Supplier QA Bio Inc Supplier No. LCSA6
S Cartridges -Fluorescent Labeling Cleanup for Glycans• One of the most reliable methods for achieving post-labeling cleanup is use of S-cartridges. Ludger S Cartridges are routinely used in the biopharma industry during glycoprofiling QC for lot release of FDA and EMA approved biopharmaceuticals.