Learn More
ACROBiosystems Epstein-Barr Virus Glycoprotein B, His Tag, 100 μg
Catalog No. 502231181
Encompass_Preferred
Epstein-Barr virus Envelope Glycoprotein B (gB), His Tag is a recombinant protein expressed in human 293 (HEK293) cells. It consists of amino acids Gln 23 to Gly 685 of the Epstein-Barr virus glycoprotein B and is designed for research applications, particularly in studies involving viral entry and membrane fusion processes. This protein is supplied in a lyophilized format.
  • Greater than 95% purity as determined by SDS-PAGE
  • Features a C-terminal polyhistidine tag
  • Validated by MALS to confirm molecular weight
  • Less than 1.0 EU per μg endotoxin level measured by LAL or rFC method
  • Expressed in mammalian HEK293 cells for proper post-translational modifications
  • Stable for 12 months in lyophilized state at -20°C to -70°C
This item is not returnable. View return policy
Catalog No. 50-223-1181 Supplier ACROBiosystems Supplier No. GLBE52H3100UG
Epstein-Barr virus Envelope Glycoprotein B (gB), His Tag is a recombinant protein expressed in human 293 (HEK293) cells. It consists of amino acids Gln 23 to Gly 685 of the Epstein-Barr virus glycoprotein B and is designed for research applications, particularly in studies involving viral entry and membrane fusion processes. This protein is supplied in a lyophilized format.
  • Greater than 95% purity as determined by SDS-PAGE
  • Features a C-terminal polyhistidine tag
  • Validated by MALS to confirm molecular weight
  • Less than 1.0 EU per μg endotoxin level measured by LAL or rFC method
  • Expressed in mammalian HEK293 cells for proper post-translational modifications
  • Stable for 12 months in lyophilized state at -20°C to -70°C