Learn More
Apexbio Technology LLC Glycoprotein B (485-492) 10mg
SDP
Catalog No. 5000095572
Encompass_Preferred
Glycoprotein B 485-492 is a viral glycoprotein-derived peptide fragment from herpes simplex virus (HSV) It is designed to facilitate virus-cell membrane fusion by interacting with cellular membranes thereby promoting HSV entry into host cells Glycoprotein B 485-492 exerts its biological activity primarily through its role as part of the viral fusion machinery in conjunction with glycoproteins H and L Based on these properties Glycoprotein B 485-492 holds research potential in investigating HSV fusion mechanisms antiviral development and antigen-antibody interaction studies in viral entry
Catalog No. 50-000-95572 Supplier Apexbio Technology LLC Supplier No. A113810
Glycoprotein B 485-492 is a viral glycoprotein-derived peptide fragment from herpes simplex virus (HSV) It is designed to facilitate virus-cell membrane fusion by interacting with cellular membranes thereby promoting HSV entry into host cells Glycoprotein B 485-492 exerts its biological activity primarily through its role as part of the viral fusion machinery in conjunction with glycoproteins H and L Based on these properties Glycoprotein B 485-492 holds research potential in investigating HSV fusion mechanisms antiviral development and antigen-antibody interaction studies in viral entry