Learn More
ACROBiosystems Glycoprotein D HSV-2 100ug
Catalog No. 502535015
Encompass_Preferred
Herpesvirus infections are widely spread throughout the world population. Herpes simplex virus (HSV) belongs to the α-herpesvirus subfamily. There are two main types of HSV HSV-1 and HSV-2 which infect humans. HSV-2 mainly causes genital lesions whereas HSV-1 is involved in both oral and genital infections. Glycoprotein D (gD) is a structural component of the herpes simplex virus type 1 (HSV-1) envelope which is essential for virus entry and fusion with host cells. gD plays an important role by binding to the host receptors such as herpes virus entry mediator (HVEM) and nectin-1 a member of the immunoglobulin (Ig)-like cell adhesion molecules.
Catalog No. 50-253-5015 Supplier ACROBiosystems Supplier No. GLDM609100UG
Herpesvirus infections are widely spread throughout the world population. Herpes simplex virus (HSV) belongs to the α-herpesvirus subfamily. There are two main types of HSV HSV-1 and HSV-2 which infect humans. HSV-2 mainly causes genital lesions whereas HSV-1 is involved in both oral and genital infections. Glycoprotein D (gD) is a structural component of the herpes simplex virus type 1 (HSV-1) envelope which is essential for virus entry and fusion with host cells. gD plays an important role by binding to the host receptors such as herpes virus entry mediator (HVEM) and nectin-1 a member of the immunoglobulin (Ig)-like cell adhesion molecules.