Learn More
ACROBiosystems Varicella zoster virus (strain Oka vaccine) Envelope Glycoprotein E (gE), His Tag (GLE-V52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 31 - Ala 546 (Accession # Q9J3M8).
Catalog No. 502231177
Encompass_Preferred
Varicella-zoster virus (VZV) is a neurotropic virus belonging to the Herpesviridae family. Primary VZV infection causes chickenpox and is followed by a life-long latent infection established mainly in the cranial and dorsal root ganglia. Reactivation of the virus is often associated with shingles (herpes zoster). Glycoprotein E (gE) is one of the known glycoproteins (gB, gC, gE, gH, gI, gK, gI) of VZV that is most abundantly expressed on the surface of virus and infected cells, playing an important role in viral replication and cell-to-cell spread. The strongly immunogenic gE can provide strong IgG signal in body fluid, which makes it ideal to be developed as an antigen for analysis of IgG antibodies. gE also demonstrates high potency as a vaccine immunogen and is formulated as the single viral envelope protein that constitutes the GSK VZV recombinant subunit vaccine Shingrix.
This item is not returnable. View return policy
Catalog No. 50-223-1177 Supplier ACROBiosystems Supplier No. GLEV52H3100UG
Varicella-zoster virus (VZV) is a neurotropic virus belonging to the Herpesviridae family. Primary VZV infection causes chickenpox and is followed by a life-long latent infection established mainly in the cranial and dorsal root ganglia. Reactivation of the virus is often associated with shingles (herpes zoster). Glycoprotein E (gE) is one of the known glycoproteins (gB, gC, gE, gH, gI, gK, gI) of VZV that is most abundantly expressed on the surface of virus and infected cells, playing an important role in viral replication and cell-to-cell spread. The strongly immunogenic gE can provide strong IgG signal in body fluid, which makes it ideal to be developed as an antigen for analysis of IgG antibodies. gE also demonstrates high potency as a vaccine immunogen and is formulated as the single viral envelope protein that constitutes the GSK VZV recombinant subunit vaccine Shingrix.