Learn More
Vector Laboratories Glycoprotein Eluting Solution for Sialic Acid-Binding Lectins 100 ml
SDP
Catalog No. NC0612573
Encompass
This Glycoprotein Eluting Solution is for eluting glycoproteins from agarose-bound sialic acid-binding lectins. The neutral pH of this solution maximizes the yield of eluted glycoproteins and preserves the activity of the agarose-bound lectins for repeated use. All components of this ready-to-use solution can be subsequently removed by dialysis. This Glycoprotein Eluting Solution offers convenience and superior recovery over standard sugar solutions.
Catalog No. NC0612573 Supplier Vector Laboratories Supplier No. ES7100100
This Glycoprotein Eluting Solution is for eluting glycoproteins from agarose-bound sialic acid-binding lectins. The neutral pH of this solution maximizes the yield of eluted glycoproteins and preserves the activity of the agarose-bound lectins for repeated use. All components of this ready-to-use solution can be subsequently removed by dialysis. This Glycoprotein Eluting Solution offers convenience and superior recovery over standard sugar solutions.