Learn More
Vector Laboratories Glycoprotein Eluting Solution for GlcNAc or Chitin-Binding Lectins 100 ml
SDP
Catalog No. NC1253589
Encompass
This Glycoprotein Eluting Solution can be used to elute glycoproteins from agarose-bound N-acetylglucosamine- or chitin-binding lectins. The neutral pH of this solution maximizes the yield of eluted glycoproteins and preserves the activity of the agarose-bound lectins for repeated use. All components of this ready-to-use solution can be subsequently removed by dialysis. This Glycoprotein Eluting Solution offers convenience and superior recovery over standard sugar solutions.
Catalog No. NC1253589 Supplier Vector Laboratories Supplier No. ES5100100
This Glycoprotein Eluting Solution can be used to elute glycoproteins from agarose-bound N-acetylglucosamine- or chitin-binding lectins. The neutral pH of this solution maximizes the yield of eluted glycoproteins and preserves the activity of the agarose-bound lectins for repeated use. All components of this ready-to-use solution can be subsequently removed by dialysis. This Glycoprotein Eluting Solution offers convenience and superior recovery over standard sugar solutions.