Learn More
ACROBiosystems ACROBiosystems Ebolavirus (subtype Bundibugyo, strain Uganda 2007) Envelope Glycoprotein 1 (GP1) Protein, His Tag
Catalog No. 501957207
Encompass_Preferred
Recombinant Protein;1MG;Ebolavirus (subtype Bundibugyo, strain Uganda 2007) GP1, His Tag (BEP-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Gln 304 (Accession # ACI28624).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.8 kDa. The protein migrates as 40-60 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7207 Supplier ACROBiosystems Supplier No. BEPV52211MG
Recombinant Protein;1MG;Ebolavirus (subtype Bundibugyo, strain Uganda 2007) GP1, His Tag (BEP-V5221) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Gln 304 (Accession # ACI28624).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.8 kDa. The protein migrates as 40-60 kDa under reducing (R) condition (SDS-PAGE).