Learn More
ACROBiosystems FITC-Labeled Rabies Virus Glycoprotein G (strain CVS-11), His tag, 25 μg (ACROBiosystems GLN-RF5E3-25UG)
Catalog No. 502357335
Encompass_Preferred
FITC-labeled recombinant Glycoprotein G from Rabies virus (strain CVS-11) with a C-terminal His tag. Expressed in baculovirus-insect cells and supplied lyophilized (25 μg); designed for use as a fluorescent antigen in immunoassays and binding studies.
  • FITC-labeled glycoprotein for fluorescence-based assays.
  • Expressed in baculovirus-insect cells.
  • C-terminal polyhistidine (His) tag.
  • Lyophilized format with trehalose protectant.
  • Purity greater than 90% by SDS-PAGE.
  • Apparent molecular weight 53.6 kDa; migrates as 60-62 kDa and >116 kDa under reducing conditions.
  • Store lyophilized at -20°C or lower; protect from light and avoid repeated freeze-thaw.
  • Routinely tested for immobilization at 1 μg/mL (100 μL/well) for binding assays.
Catalog No. 50-235-7335 Supplier ACROBiosystems Supplier No. GLNRF5E325UG
FITC-labeled recombinant Glycoprotein G from Rabies virus (strain CVS-11) with a C-terminal His tag. Expressed in baculovirus-insect cells and supplied lyophilized (25 μg); designed for use as a fluorescent antigen in immunoassays and binding studies.
  • FITC-labeled glycoprotein for fluorescence-based assays.
  • Expressed in baculovirus-insect cells.
  • C-terminal polyhistidine (His) tag.
  • Lyophilized format with trehalose protectant.
  • Purity greater than 90% by SDS-PAGE.
  • Apparent molecular weight 53.6 kDa; migrates as 60-62 kDa and >116 kDa under reducing conditions.
  • Store lyophilized at -20°C or lower; protect from light and avoid repeated freeze-thaw.
  • Routinely tested for immobilization at 1 μg/mL (100 μL/well) for binding assays.