Learn More
ACROBiosystems Mouse Anti-RABV Glycoprotein IgG Titer Serologic Assay Kit (ELISA), 96 tests
Catalog No. 502370194
Encompass_Preferred
Serologic ELISA kit for measuring anti-RABV (rabies virus) glycoprotein IgG titers in mouse serum. Designed for preclinical vaccine immunogenicity evaluation, the kit contains a pre-coated microplate, positive and negative controls, HRP-conjugated antibody, buffers, substrate, and stop solution for 96 tests. Store unopened at 2 °C-8 °C; follow component-specific storage after opening; check the external packaging for expiration date.
  • Measures anti-RABV glycoprotein IgG titers in mouse serum
  • Indirect ELISA format for quantitative antibody titer assessment
  • Includes pre-coated microplate and positive and negative controls
  • Contains HRP-conjugated antibody, washing buffer, dilution buffer, substrate, and stop solution
  • Suitable for 96 tests per kit
  • Unopened storage 2 °C-8 °C; 30-day shelf life after opening
Catalog No. 50-237-0194 Supplier ACROBiosystems Supplier No. RAST15796TESTS
Serologic ELISA kit for measuring anti-RABV (rabies virus) glycoprotein IgG titers in mouse serum. Designed for preclinical vaccine immunogenicity evaluation, the kit contains a pre-coated microplate, positive and negative controls, HRP-conjugated antibody, buffers, substrate, and stop solution for 96 tests. Store unopened at 2 °C-8 °C; follow component-specific storage after opening; check the external packaging for expiration date.
  • Measures anti-RABV glycoprotein IgG titers in mouse serum
  • Indirect ELISA format for quantitative antibody titer assessment
  • Includes pre-coated microplate and positive and negative controls
  • Contains HRP-conjugated antibody, washing buffer, dilution buffer, substrate, and stop solution
  • Suitable for 96 tests per kit
  • Unopened storage 2 °C-8 °C; 30-day shelf life after opening