Learn More
Bioworld Glycoprotein Staining Kit, 1 Kit
Catalog No. 50198591
Encompass_Preferred
Glycoprotein Staining KitIncorporates the proven Schiff's reagent and method.Specifications:One kit contains enough reagents to stain 25 mini gels Kit Contains: Oxidation Reagent Reduction Reagent Schiff's Reagent Glycoprotein Standard
Catalog No. 50-198-591 Supplier Bioworld Supplier No. 213400011
Glycoprotein Staining KitIncorporates the proven Schiff's reagent and method.Specifications:One kit contains enough reagents to stain 25 mini gels Kit Contains: Oxidation Reagent Reduction Reagent Schiff's Reagent Glycoprotein Standard