Learn More
ACROBiosystems Recombinant Protein;Recombinant HCMV Glycoprotein B (gB), Fc Tag;HEK293;500UG;CMB,CMV,HCMV,GB,Envelope glycoprotein B,Glycoprotein GP55,UL55,GP55
Catalog No. 501658614
Encompass_Preferred
Recombinant Protein;Recombinant HCMV Glycoprotein B (gB), Fc Tag;500UG;Recombinant Human cytomegalovirus (HCMV) Glycoprotein B /gB Protein, C-Fc Tag (rh CMV-GB Fc Chimera) Val 23 - Lys 700 (Accession # AAA45920.1) with furin cleavage site mutated from 'RTKR' to 'TTQT', was produced in human 293 cells (HEK293) at ACROBiosystems.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104 kDa. The protein migrates as 140-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8614 Supplier ACROBiosystems Supplier No. CMBV5255500UG
Recombinant Protein;Recombinant HCMV Glycoprotein B (gB), Fc Tag;500UG;Recombinant Human cytomegalovirus (HCMV) Glycoprotein B /gB Protein, C-Fc Tag (rh CMV-GB Fc Chimera) Val 23 - Lys 700 (Accession # AAA45920.1) with furin cleavage site mutated from 'RTKR' to 'TTQT', was produced in human 293 cells (HEK293) at ACROBiosystems.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 104 kDa. The protein migrates as 140-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.