Learn More
ACROBiosystems Recombinant Protein;Recombinant Ebolavirus EBOV (subtype Zaire,strain Kikwit-95) Envelope Glycoprotein 1 (GP1);HEK293;100UG;ZE5,CMV,HCMV,GB,Envelope glycoprotein B,Glycoprotein GP55,UL55,GP55
Catalog No. 501658620
Encompass_Preferred
Recombinant Protein;Recombinant Ebolavirus EBOV (subtype Zaire,strain Kikwit-95) Envelope Glycoprotein 1 (GP1);100UG;Ebolavirus GP (virus), His Tag (ZE5-V5220) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Arg 501 (Accession # AAQ55048.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.6 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8620 Supplier ACROBiosystems Supplier No. ZE5V5220100UG
Recombinant Protein;Recombinant Ebolavirus EBOV (subtype Zaire,strain Kikwit-95) Envelope Glycoprotein 1 (GP1);100UG;Ebolavirus GP (virus), His Tag (ZE5-V5220) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Arg 501 (Accession # AAQ55048.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.6 kDa. The protein migrates as 120-140 kDa under reducing (R) condition (SDS-PAGE).