Learn More
ACROBiosystems Recombinant Protein;Recombinant Ebolavirus BDBV(subtype Bundibugyo,strain Uganda 2007) Small/secreted Glycoprotein (sGP);HEK293;1MG;BEP,CMV,HCMV,GB,Envelope glycoprotein B,Glycoprotein GP55,UL55,GP55
Supplier:  ACROBiosystems BEPV52201MG
Encompass_Preferred
Recombinant Protein;Recombinant Ebolavirus BDBV(subtype Bundibugyo,strain Uganda 2007) Small/secreted Glycoprotein (sGP);1MG;Ebolavirus GP (virus), His Tag (BEP-V5220) is expressed from human 293 cells (HEK293). It contains AA Ile 33 - Arg 324 (Accession # B8XCN1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.3 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8631