Learn More
Apexbio Technology LLC Glycopyrrolate 596-51-0 10mg
SDP
Catalog No. 502240454
Encompass_Preferred
Glycopyrrolate (CAS 596-51-0) is a small molecule that acts as a competitive antagonist of muscarinic acetylcholine receptors By blocking muscarinic receptor-mediated cholinergic signaling it inhibits smooth muscle contractions and reduces secretory gland activity This pharmacological profile makes glycopyrrolate a valuable tool in studies investigating muscarinic receptor function neurotransmission and cholinergic pathways It is frequently used in research focused on gastrointestinal motility airway regulation and autonomic nervous system modulation
This item is not returnable. View return policy
Catalog No. 50-224-0454 Supplier Apexbio Technology LLC Supplier No. B126610
Glycopyrrolate (CAS 596-51-0) is a small molecule that acts as a competitive antagonist of muscarinic acetylcholine receptors By blocking muscarinic receptor-mediated cholinergic signaling it inhibits smooth muscle contractions and reduces secretory gland activity This pharmacological profile makes glycopyrrolate a valuable tool in studies investigating muscarinic receptor function neurotransmission and cholinergic pathways It is frequently used in research focused on gastrointestinal motility airway regulation and autonomic nervous system modulation