Learn More
Medchemexpress LLC Glycopyrrolate | 596-51-0 | 99.93% | 398.33 | 25MG
SDP
Catalog No. 5000383936
Encompass_Preferred
Glycopyrrolate (Glycopyrronium bromide) is a quaternary ammonium derivative and a muscarinic receptor antagonist. It has a bronchoprotective effect and produces a beneficial effect on blood pressure. It can be used for the research of bronchial diseases.
  • Functions as a muscarinic receptor antagonist.
  • Exhibits a bronchoprotective effect.
  • Produces a beneficial effect on blood pressure.
  • Can be utilized for research related to bronchial diseases.
Catalog No. 50-003-83936 Supplier Medchemexpress LLC Supplier No. 25MGHY17465
Glycopyrrolate (Glycopyrronium bromide) is a quaternary ammonium derivative and a muscarinic receptor antagonist. It has a bronchoprotective effect and produces a beneficial effect on blood pressure. It can be used for the research of bronchial diseases.
  • Functions as a muscarinic receptor antagonist.
  • Exhibits a bronchoprotective effect.
  • Produces a beneficial effect on blood pressure.
  • Can be utilized for research related to bronchial diseases.