Learn More
Medchemexpress LLC Glycopyrrolate iodide-d3 (mixture of diastereomers) | 873295-32-0 | MFCD09952213 | >95.0% | 448.35 g/mol | C19H25D3INO3 | 1 MG
SDP
Catalog No. 5000284313
Encompass_Preferred
Glycopyrrolate iodide-d3 (mixture of diastereomers) is a deuterium-labeled isotopic analogue of glycopyrrolate supplied as the iodide salt in small research quantities. It is intended for use as an internal standard or analytical reference in biochemical, pharmacokinetic, and mass spectrometry applications. The material is supplied as a d3-labeled mixture of diastereomers at research-grade purity.
  • Deuterium-labeled (d3) isotopologue for mass spectrometry and isotope-dilution assays.
  • Iodide salt supplied as a mixture of diastereomers.
  • Research quantity suitable for standard preparation and method development.
  • Purity typically >95% (HPLC) as reported by isotope standard suppliers.
  • Intended for use as an internal standard in quantitative analytical workflows.
  • Store and handle according to the product safety data sheet.
Catalog No. 50-002-84313 Supplier Medchemexpress LLC Supplier No. HYW6542091MG
Glycopyrrolate iodide-d3 (mixture of diastereomers) is a deuterium-labeled isotopic analogue of glycopyrrolate supplied as the iodide salt in small research quantities. It is intended for use as an internal standard or analytical reference in biochemical, pharmacokinetic, and mass spectrometry applications. The material is supplied as a d3-labeled mixture of diastereomers at research-grade purity.
  • Deuterium-labeled (d3) isotopologue for mass spectrometry and isotope-dilution assays.
  • Iodide salt supplied as a mixture of diastereomers.
  • Research quantity suitable for standard preparation and method development.
  • Purity typically >95% (HPLC) as reported by isotope standard suppliers.
  • Intended for use as an internal standard in quantitative analytical workflows.
  • Store and handle according to the product safety data sheet.