Learn More
LECTENZ BIO INC GlycoSense™ Array Kit 50ul
SDP
Catalog No. NC1910500
Encompass_Preferred
GlycoSense™ is a multiplex assay for the detection of glycosylation patterns on glycoproteins using a two-laser flow cytometer. The GlycoSense™ array consists of lectins conjugated to multiplex microspheres (beads) that bind terminal monosaccharides on a glycoprotein to quickly profile the glycan features present. This simplified “GlycoPrint” does not give a full glycosylation profile or structural information, however, is capable of detecting changes in several key glycan features. The microspheres in this array have been evaluated to bind sialic acid, galactose and N-acetylglucosamine (GlcNAc) on N-linked glycans. LECTENZ BIO mfg# GK0101-50UL
Catalog No. NC1910500 Supplier LECTENZ BIO INC Supplier No. GK010150UL
GlycoSense™ is a multiplex assay for the detection of glycosylation patterns on glycoproteins using a two-laser flow cytometer. The GlycoSense™ array consists of lectins conjugated to multiplex microspheres (beads) that bind terminal monosaccharides on a glycoprotein to quickly profile the glycan features present. This simplified “GlycoPrint” does not give a full glycosylation profile or structural information, however, is capable of detecting changes in several key glycan features. The microspheres in this array have been evaluated to bind sialic acid, galactose and N-acetylglucosamine (GlcNAc) on N-linked glycans. LECTENZ BIO mfg# GK0101-50UL